Learn More
Medchemexpress LLC Encequidar mesylate | 849675-87-2 | MFCD25976626 | 99.8% | 784.8 g/mol | C39H40N6O10S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY13646A5MG
Encompass_Preferred
Encequidar mesylate is the mesylate salt of encequidar, a potent, competitive inhibitor of P-glycoprotein (P-gp/ABCB1) used as a research tool to probe transporter-mediated drug disposition, oral bioavailability, and drug-drug interactions. It is provided as a high-purity solid suitable for in vitro and in vivo studies.
  • Potent competitive inhibitor of P-glycoprotein for transporter studies.
  • High reported purity (99.82%), suitable for analytical and biological work.
  • Available in small pre-weighed quantities for dosing and assay use.
  • Provides molecular formula and molecular weight for formulation and calculation.
  • Supplied as a mesylate salt to improve stability and handling.
Catalog No. 50-193-2672