Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Encequidar mesylate | 849675-87-2 | 99.6% | 784.83 | 50 MG
SDP

Catalog No. 5000350313
Encompass_Preferred

Encequidar mesylate is a competitive and potent P-glycoprotein inhibitor. This product is for research use only.

  • Target: P-glycoprotein
  • Appearance: Solid, light yellow to yellow
  • Solubility in DMSO: 25 mg/mL (31.85 mM), requires ultrasonic
  • Improved in vivo absorption with microcapsule form
  • Involved in clinical trials for metastatic breast cancer, breast neoplasms, and solid tumors

Catalog No. 50-003-50313 Supplier Medchemexpress LLC Supplier No. HY13646A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Encequidar mesylate is a competitive and potent P-glycoprotein inhibitor. This product is for research use only.

  • Target: P-glycoprotein
  • Appearance: Solid, light yellow to yellow
  • Solubility in DMSO: 25 mg/mL (31.85 mM), requires ultrasonic
  • Improved in vivo absorption with microcapsule form
  • Involved in clinical trials for metastatic breast cancer, breast neoplasms, and solid tumors

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.