Learn More
Medchemexpress LLC Encequidar mesylate | 849675-87-2 | 99.8% | 784.83 | 100 MG
SDP
Catalog No. 5000351616
Encompass_Preferred
Encequidar mesylate is a competitive and potent P-glycoprotein inhibitor intended for research use only. It has demonstrated significant inhibition of survival in in vitro studies and improved absorption in vivo when administered in microcapsule form due to enhanced aqueous solubility and dissolution.
  • Competitive and potent p-glycoprotein inhibitor
  • Enhances absorption through improved aqueous solubility
  • Provides faster Tmax and higher AUC values in microcapsule form
  • Demonstrates inhibition of survival in in vitro studies
Catalog No. 50-003-51616 Supplier Medchemexpress LLC Supplier No. HY13646A100MG
Encequidar mesylate is a competitive and potent P-glycoprotein inhibitor intended for research use only. It has demonstrated significant inhibition of survival in in vitro studies and improved absorption in vivo when administered in microcapsule form due to enhanced aqueous solubility and dissolution.
  • Competitive and potent p-glycoprotein inhibitor
  • Enhances absorption through improved aqueous solubility
  • Provides faster Tmax and higher AUC values in microcapsule form
  • Demonstrates inhibition of survival in in vitro studies