Learn More
Cayman Chemical Enclomiphene (citrate), Purity: ≥95%, CAS: 7599-79-3, 10 mg
SDP
Catalog No. 502989545
Encompass_Preferred
A selective estrogen receptor modulator; can act as an ER antagonist or agonist in a dose-, tissue-, and species-dependent manner; decreases or increases serum testosterone levels in male rats or baboons, respectively
Catalog No. 50-298-9545 Supplier Cayman Chemical Supplier No. 3096510 MG
A selective estrogen receptor modulator; can act as an ER antagonist or agonist in a dose-, tissue-, and species-dependent manner; decreases or increases serum testosterone levels in male rats or baboons, respectively