Learn More
Medchemexpress LLC HY-118861A 10mg Medchemexpress, Enclomiphene (citrate) CAS:7599-79-3 Purity:>98%
SDP
Catalog No. 501969511
Encompass_Preferred
Medchemexpress, HY-118861A 10mg Enclomiphene (citrate) CAS:7599-79-3 Enclomiphene citrate is a potent and orally active oestrogen receptor antagonist, with antioestrogenic property. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9511 Supplier Medchemexpress LLC Supplier No. HY118861A10MG
Medchemexpress, HY-118861A 10mg Enclomiphene (citrate) CAS:7599-79-3 Enclomiphene citrate is a potent and orally active oestrogen receptor antagonist, with antioestrogenic property. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.