Learn More
Selleck Chemical LLC Enclomiphene Citrate
SDP
Catalog No. 503138365
Encompass_Preferred
Enclomiphene citrate is the trans isomer of clomiphene citrate and acts as a non-steroidal estrogen receptor antagonist
Catalog No. 50-313-8365 Supplier Selleck Chemical LLC Supplier No. E0361100MG
Enclomiphene citrate is the trans isomer of clomiphene citrate and acts as a non-steroidal estrogen receptor antagonist