Learn More
Medchemexpress LLC Enclomiphene citrate | 7599-79-3 | 100.0% | 598.09 | C32H36ClNO8 | 5 MG
SDP
Catalog No. 5000228622
Encompass_Preferred
Enclomiphene citrate is a research-grade, non-steroidal estrogen receptor antagonist (the (E)-isomer of clomiphene) used in studies of ovarian dysfunction, testosterone deficiency, male hypogonadism, and type 2 diabetes. It is supplied as a small quantity for laboratory research and assay development.
  • Research-grade estrogen receptor antagonist with antioestrogenic properties.
  • Used in studies of ovarian dysfunction, hormonal regulation, and metabolic disease.
  • Suitable for in vitro screening and early-stage pharmacology research.
  • Compact sample size for assay development and method validation.
  • Molecular formula C32H36ClNO8; molecular weight 598.09.
Catalog No. 50-002-28622 Supplier Medchemexpress LLC Supplier No. HY118861A5MG
Enclomiphene citrate is a research-grade, non-steroidal estrogen receptor antagonist (the (E)-isomer of clomiphene) used in studies of ovarian dysfunction, testosterone deficiency, male hypogonadism, and type 2 diabetes. It is supplied as a small quantity for laboratory research and assay development.
  • Research-grade estrogen receptor antagonist with antioestrogenic properties.
  • Used in studies of ovarian dysfunction, hormonal regulation, and metabolic disease.
  • Suitable for in vitro screening and early-stage pharmacology research.
  • Compact sample size for assay development and method validation.
  • Molecular formula C32H36ClNO8; molecular weight 598.09.