Learn More
Apexbio Technology LLC Enclomiphene citrate 7599-79-3 5mg
SDP
Catalog No. 5000096024
Encompass_Preferred
Enclomiphene citrate (CAS 7599-79-3) is a small-molecule antagonist targeting estrogen receptors It is designed to competitively bind to estrogen receptors thereby inhibiting receptor-mediated estrogen signaling and attenuating estrogen-induced negative feedback on the hypothalamic-pituitary axis Enclomiphene citrate exerts its biological activity primarily by increasing endogenous secretion of gonadotropins including luteinizing hormone (LH) and follicle stimulating hormone (FSH) which promotes endogenous testosterone biosynthesis in gonadal tissues Based on these pharmacological properties enclomiphene citrate holds research potential in studies of male hypogonadism fertility disorders and regulation of male gonadal function
Catalog No. 50-000-96024 Supplier Apexbio Technology LLC Supplier No. A99745
Enclomiphene citrate (CAS 7599-79-3) is a small-molecule antagonist targeting estrogen receptors It is designed to competitively bind to estrogen receptors thereby inhibiting receptor-mediated estrogen signaling and attenuating estrogen-induced negative feedback on the hypothalamic-pituitary axis Enclomiphene citrate exerts its biological activity primarily by increasing endogenous secretion of gonadotropins including luteinizing hormone (LH) and follicle stimulating hormone (FSH) which promotes endogenous testosterone biosynthesis in gonadal tissues Based on these pharmacological properties enclomiphene citrate holds research potential in studies of male hypogonadism fertility disorders and regulation of male gonadal function