Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Enclomiphene citrate | 7599-79-3 | 99.9% | 598.09 | C32H36ClNO8 | 1 ML
SDP

Catalog No. 5000229862
Encompass_Preferred

Enclomiphene citrate is a research-grade, non-steroidal estrogen receptor antagonist supplied as a solid or as a ready-to-use 10 mM solution in DMSO for in vitro and in vivo studies.

  • High purity typically 99.93%.
  • Available as a 10 mM solution (1 mL) in DMSO or as mg-scale solids.
  • Used for studies of ovarian dysfunction, testosterone deficiency, and male hypogonadism.
  • Molecular weight 598.09, formula C32H36ClNO8, CAS 7599-79-3.
  • Store powder at 4°C; stock solutions at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-29862 Supplier Medchemexpress LLC Supplier No. HY118861A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Enclomiphene citrate is a research-grade, non-steroidal estrogen receptor antagonist supplied as a solid or as a ready-to-use 10 mM solution in DMSO for in vitro and in vivo studies.

  • High purity typically 99.93%.
  • Available as a 10 mM solution (1 mL) in DMSO or as mg-scale solids.
  • Used for studies of ovarian dysfunction, testosterone deficiency, and male hypogonadism.
  • Molecular weight 598.09, formula C32H36ClNO8, CAS 7599-79-3.
  • Store powder at 4°C; stock solutions at -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.