Learn More
Medchemexpress LLC Enclomiphene-d4 | 00-00-0 | 99.9% | 409.98 g/mol | C26H24D4ClNO | 5 MG
SDP
Catalog No. 5000264368
Encompass_Preferred
Enclomiphene-d4 is a deuterium-labeled analogue of enclomiphene provided for use as a tracer and internal standard in quantitative analytical workflows. It is intended for research applications where stable-isotope labeling improves accuracy and precision in NMR, GC-MS, and LC-MS assays.
  • Deuterium-labeled internal standard for quantitative analysis by NMR, GC-MS, and LC-MS.
  • High purity: 99.85%.
  • Molecular formula C26H24D4ClNO and molecular weight 409.98 g/mol.
  • Available in small research quantities suitable for analytical workflows.
  • Includes supporting documentation such as COA, HNMR, RP-HPLC, and LCMS reports.
Catalog No. 50-002-64368 Supplier Medchemexpress LLC Supplier No. HY118861S15MG
Enclomiphene-d4 is a deuterium-labeled analogue of enclomiphene provided for use as a tracer and internal standard in quantitative analytical workflows. It is intended for research applications where stable-isotope labeling improves accuracy and precision in NMR, GC-MS, and LC-MS assays.
  • Deuterium-labeled internal standard for quantitative analysis by NMR, GC-MS, and LC-MS.
  • High purity: 99.85%.
  • Molecular formula C26H24D4ClNO and molecular weight 409.98 g/mol.
  • Available in small research quantities suitable for analytical workflows.
  • Includes supporting documentation such as COA, HNMR, RP-HPLC, and LCMS reports.