Learn More
Medchemexpress LLC Enclomiphene-d4 | 15690-57-0 | 99.85% | 409.98 | 10 MG
SDP
Catalog No. 5000436197
Encompass_Preferred
Enclomiphene-d4 is a deuterium labeled Enclomiphene. Enclomiphene is a potent and orally active estrogen receptor antagonist with antioestrogenic property. It is for research use only.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-36197 Supplier Medchemexpress LLC Supplier No. HY118861S110MG
Enclomiphene-d4 is a deuterium labeled Enclomiphene. Enclomiphene is a potent and orally active estrogen receptor antagonist with antioestrogenic property. It is for research use only.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.