Learn More
Medchemexpress LLC Enclomiphene-d4 hydrochloride | 00-00-0 | MFCD00867469 | 99.0% | 446.45 g/mol | C26H25D4Cl2NO | 5 MG
SDP
Catalog No. 5000264834
Encompass_Preferred
Enclomiphene-d4 hydrochloride is a deuterium-labeled analogue of enclomiphene supplied as the hydrochloride salt for research use. It is intended primarily as an internal standard or tracer in analytical and bioanalytical workflows, supporting accurate quantitation and method validation in LC-MS, GC-MS, and NMR applications.
  • Isotopically labeled for use as an internal standard in mass spectrometry.
  • High purity supports reliable analytical results.
  • Provided as a hydrochloride salt for consistent physicochemical properties.
  • Suitable for method development, validation, and quantitative assays.
  • Available in small milligram pack sizes for laboratory use.
Catalog No. 50-002-64834 Supplier Medchemexpress LLC Supplier No. HY118861S5MG
Enclomiphene-d4 hydrochloride is a deuterium-labeled analogue of enclomiphene supplied as the hydrochloride salt for research use. It is intended primarily as an internal standard or tracer in analytical and bioanalytical workflows, supporting accurate quantitation and method validation in LC-MS, GC-MS, and NMR applications.
  • Isotopically labeled for use as an internal standard in mass spectrometry.
  • High purity supports reliable analytical results.
  • Provided as a hydrochloride salt for consistent physicochemical properties.
  • Suitable for method development, validation, and quantitative assays.
  • Available in small milligram pack sizes for laboratory use.