Learn More
Medchemexpress LLC Enclomiphene-d4 hydrochloride | 15690-57-0 | 99.0% | 446.45 | 1 MG
SDP
Catalog No. 5000437792
Encompass_Preferred
Enclomiphene-d4 hydrochloride is a deuterium labeled Enclomiphene hydrochloride. It is a potent and orally active estrogen receptor antagonist with antioestrogenic property.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-37792 Supplier Medchemexpress LLC Supplier No. HY118861S1MG
Enclomiphene-d4 hydrochloride is a deuterium labeled Enclomiphene hydrochloride. It is a potent and orally active estrogen receptor antagonist with antioestrogenic property.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.