Learn More
Medchemexpress LLC Enclomiphene-d4 hydrochloride | 15690-57-0 | 99.0% | 446.45 | 10 MG
SDP
Catalog No. 5000437793
Encompass_Preferred
Enclomiphene-d4 hydrochloride is a deuterium labeled Enclomiphene hydrochloride. It is a potent and orally active estrogen receptor antagonist with antioestrogenic property. Deuteration has garnered attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • For research use only.
  • Stable heavy isotopes incorporated into drug molecules for quantitation during drug development.
Catalog No. 50-004-37793 Supplier Medchemexpress LLC Supplier No. HY118861S10MG
Enclomiphene-d4 hydrochloride is a deuterium labeled Enclomiphene hydrochloride. It is a potent and orally active estrogen receptor antagonist with antioestrogenic property. Deuteration has garnered attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • For research use only.
  • Stable heavy isotopes incorporated into drug molecules for quantitation during drug development.