Learn More
Grainger METRO Enclosure Panel, Steel, Chrome Finish, 18-3/8 in. x 59-3/4 in., 1 ea
Catalog No. 502779670
Encompass_Preferred
These steel enclosure panels are designed for use with wire shelving units to provide containment for stored items. The panels attach to the back or sides of stationary and mobile shelving, preventing items from falling off and securing the workspace.
  • Durable steel construction with a chrome finish
  • Includes necessary mounting hardware for installation
  • Compatible with 63 in. high posts
  • Helps prevent items from falling off shelf edges
  • Grid design for clear visibility of stored contents
Catalog No. 50-277-9670 Supplier Grainger Supplier No. 19N116
These steel enclosure panels are designed for use with wire shelving units to provide containment for stored items. The panels attach to the back or sides of stationary and mobile shelving, preventing items from falling off and securing the workspace.
  • Durable steel construction with a chrome finish
  • Includes necessary mounting hardware for installation
  • Compatible with 63 in. high posts
  • Helps prevent items from falling off shelf edges
  • Grid design for clear visibility of stored contents