Learn More
MSC Electra-Kool Filtered Enclosure Blower, 60 CFM, 3030 RPM, 1 ea
Catalog No. 5000503702
Encompass
The Electra-Kool filtered enclosure blower is designed to maintain optimal operating temperatures within industrial enclosures by providing efficient airflow. It is engineered for single-phase operation, delivering a consistent air movement to prevent overheating of sensitive electronic components.
  • Operates on 1 phase
  • Has 0.49 amp current draw
  • Features 3030 RPM motor speed
  • Provides 60 CFM airflow
  • Is filtered to protect internal components
Catalog No. 50-005-03702 Supplier MSC Supplier No. 03561313
The Electra-Kool filtered enclosure blower is designed to maintain optimal operating temperatures within industrial enclosures by providing efficient airflow. It is engineered for single-phase operation, delivering a consistent air movement to prevent overheating of sensitive electronic components.
  • Operates on 1 phase
  • Has 0.49 amp current draw
  • Features 3030 RPM motor speed
  • Provides 60 CFM airflow
  • Is filtered to protect internal components