Learn More
BRANSON ULTRASONICS CORPORATION (3P) ENCLOSURE SOUND PROOF
Catalog No. NC3721571
Encompass_Preferred
Branson Ultrasonics Soundproof Enclosure, For Use With: Sonifier cell disruptor, Ideal for minimizes mechanical noise levels by 20 to 25dBA produced by ultrasonic processing, Dimensions: 12.5 x 18.5 x 28 in. (31.8 x 47 x 71.1cm), Height: 28 in., 71.1cm, Length: 12.5 in., 31.8cm
Catalog No. NC3721571 Supplier BRANSON ULTRASONICS CORPORATION (3P) Supplier No. 101063275
Branson Ultrasonics Soundproof Enclosure, For Use With: Sonifier cell disruptor, Ideal for minimizes mechanical noise levels by 20 to 25dBA produced by ultrasonic processing, Dimensions: 12.5 x 18.5 x 28 in. (31.8 x 47 x 71.1cm), Height: 28 in., 71.1cm, Length: 12.5 in., 31.8cm