Learn More
Vernier™ Encoder Fan Cart
SDPSCI_ED
Catalog No. S16112ND Shop All Vernier Software Products
Change view
Click to view available options
Product Type:
Cart
Catalog No. Product Type
S16112ND Cart
1 options
Catalog No. S16112ND Supplier Vernier™ Supplier No. CARTFEC
Add to Cart
Add to Cart
Includes
Encoder Fan Cart, Motion Encoder Transmitter, Mass bars (2), Sail, User manual, The Encoder Fan Cart requires four AA batteries (not included) and the Motion Encoder Transmitter requires two AAA batteries (not included)
Requires
For use with Vernier Motion Encoder System
The Encoder Fan Cart is a large fan on a light-weight cart. It offers students the ability to perform kinematics and dynamics experiments with constant acceleration, variable mass, variable thrust, and variable thrust angle. The cart includes a Motion Encoder Transmitter for use with the Motion Encoder System.
The Encoder Fan Cart requires four AA batteries and two AAA batteries, which are not included. Either primary or rechargeable batteries can be used.
Warranty and Services
5-year limited warranty
Specifications
Product Type Cart
For Use With (Application) Educational use only
For Use With (Equipment) Dynamics Cart and Track System with Motion Encoder, Motion Encoder System
Includes Motion encoder transmitter, mass bars, sail, user manual
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov