Learn More
MSC Encon Safety Goggle, 500 Series, Clear Lens, ENFOG, 1 ea
Catalog No. 5000766334
Encompass
The Encon 500 Series Goggle is designed for high-performance eye protection. It features a lightweight body and distortion-free optics, offering superior chemical splash and impact resistance for various industrial and laboratory applications.
  • High tech, lightweight body design
  • Distortion-free optics
  • Superior chemical splash protection
  • Impact resistant
  • Replaceable lens
  • Flexible fitting
  • Provides 99.9% UVA & UVB protection to 385Nm
  • Indirect venting
Catalog No. 50-007-66334 Supplier MSC Supplier No. 93618676
The Encon 500 Series Goggle is designed for high-performance eye protection. It features a lightweight body and distortion-free optics, offering superior chemical splash and impact resistance for various industrial and laboratory applications.
  • High tech, lightweight body design
  • Distortion-free optics
  • Superior chemical splash protection
  • Impact resistant
  • Replaceable lens
  • Flexible fitting
  • Provides 99.9% UVA & UVB protection to 385Nm
  • Indirect venting