Learn More
MSC Encon Safety Products Safety Glasses, Magnifying, Anti-Fog, 1.5 Diopter
Catalog No. 5000769174
Encompass
These safety glasses from Encon Safety Products are designed for enhanced vision and protection, featuring magnifying lenses for detailed work and robust coatings for clarity and durability. They are suitable for laboratory and industrial environments requiring reliable eye protection.
  • Anti-fog coatings improve visibility under hot and humid conditions
  • Scratch-resistant lenses for extended durability
  • Full-framed design provides comprehensive eye protection
  • Non-adjustable temples ensure a secure and comfortable fit
  • Provides UV protection
  • Meets ANSI Z87.1+ standards for impact resistance
  • Magnifying lens with +1.5 diopter for precision tasks
  • Made from polycarbonate lens material for high impact resistance
Catalog No. 50-007-69174 Supplier MSC Supplier No. 06076483
These safety glasses from Encon Safety Products are designed for enhanced vision and protection, featuring magnifying lenses for detailed work and robust coatings for clarity and durability. They are suitable for laboratory and industrial environments requiring reliable eye protection.
  • Anti-fog coatings improve visibility under hot and humid conditions
  • Scratch-resistant lenses for extended durability
  • Full-framed design provides comprehensive eye protection
  • Non-adjustable temples ensure a secure and comfortable fit
  • Provides UV protection
  • Meets ANSI Z87.1+ standards for impact resistance
  • Magnifying lens with +1.5 diopter for precision tasks
  • Made from polycarbonate lens material for high impact resistance