Learn More
Medchemexpress LLC Encorafenib (Synonyms: LGX818) 10 mM * 1 mL in DMSO
SDP
Supplier:  Medchemexpress LLC HY1560510MM 1ML DMSO
Encompass_Preferred
Encorafenib (LGX818) is a highly potent BRAF inhibitor with selective anti-proliferative and apoptotic activity in cells expressing BRAFV600E (EC50=4 nM).
Catalog No. NC2888785