Learn More
Medchemexpress LLC HY-15605 100mg , Encorafenib CAS:1269440-17-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY15605100MG
Encompass_Preferred
HY-15605 100mg Encorafenib CAS: 1269440-17-6 Encorafenib (LGX818) is a highly potent BRAF inhibitor with selective anti-proliferative and apoptotic activity in cells expressing BRAFV600E (EC50=4 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0172