Learn More
Selleck Chemical LLC Encorafenib
SDP
Catalog No. 5000000998
Encompass_Preferred
Encorafenib is a highly potent RAF inhibitor with selective anti-proliferative and apoptotic activity in cells expressing B-RAF(V600E) with EC50 of 4 nM Phase 3
Catalog No. 50-000-00998 Supplier Selleck Chemical LLC Supplier No. S710825MG
Encorafenib is a highly potent RAF inhibitor with selective anti-proliferative and apoptotic activity in cells expressing B-RAF(V600E) with EC50 of 4 nM Phase 3