Learn More
Selleck Chemical LLC Encorafenib (LGX818) 5mg 1269440-17-6
SDP
Catalog No. 501364416
Encompass_Preferred
Encorafenib (LGX818) is a highly potent RAF inhibitor with selective anti-proliferative and apoptotic activity in cells expressing B-RAF(V600E) with EC50 of 4 nM. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4416 Supplier Selleck Chemical LLC Supplier No. S71085MG
Encorafenib (LGX818) is a highly potent RAF inhibitor with selective anti-proliferative and apoptotic activity in cells expressing B-RAF(V600E) with EC50 of 4 nM. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.