Learn More
Grainger End Cap 20 mmx3 mmx40 mm
Catalog No. 502766620
Encompass_Preferred
The 80/20 End Cap for 20 Series Framing is a black anodized aluminum end cap that fits 20 series 80/20 aluminum extrusions. It has 4 set screw holes to securely fasten the end cap to the extrusion. The end cap measures 2" x 2" and has a low profile sleek design to provide a finished look. Made in the USA, it can be used for creating enclosures, workstations, guarding and more using 80/20's modular aluminum framing system.
Catalog No. 50-276-6620 Supplier Grainger Supplier No. 16U355
The 80/20 End Cap for 20 Series Framing is a black anodized aluminum end cap that fits 20 series 80/20 aluminum extrusions. It has 4 set screw holes to securely fasten the end cap to the extrusion. The end cap measures 2" x 2" and has a low profile sleek design to provide a finished look. Made in the USA, it can be used for creating enclosures, workstations, guarding and more using 80/20's modular aluminum framing system.