Learn More
Perkin Elmer US LLC Concentric Nebulizer/Scott Spray Chamber End Cap Assembly for Avio 200/500 Optima 2x00/4x00/5x00/7x00 DV/8x00
Catalog No. 509043653
Encompass_Preferred
This assembly includes: Nebulizer O-Ring, Qty. 1 (00473194) Knurled Screw, Qty. 2 (00473539) Insertion Ring, Qty. 1 (00473543) O-Ring, 34.29 mm I.D., Qty. 1 (09902033) Nut, Qty. 1 (09920521) End Cap, Qty. 1 (N0681686) End Cap Washer, Qty. 1 (N0681687)
Catalog No. 50-904-3653 Supplier Perkin Elmer US LLC Supplier No. N0680504
This assembly includes: Nebulizer O-Ring, Qty. 1 (00473194) Knurled Screw, Qty. 2 (00473539) Insertion Ring, Qty. 1 (00473543) O-Ring, 34.29 mm I.D., Qty. 1 (09902033) Nut, Qty. 1 (09920521) End Cap, Qty. 1 (N0681686) End Cap Washer, Qty. 1 (N0681687)