Learn More
Waters Corp End Connector Kit, 3.9 mm/4.6 mm I.D., 40 MPa
Catalog No. 50785425 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Waters End Connector Kit is designed for use with 3.9 mm or 4.6 mm I.D. cartridges. It allows for the replacement of the cartridge while reusing the end-fittings. This kit has a maximum pressure rating of 40 MPa.
  • Can be used with any 3.9 mm or 4.6 mm I.D. cartridge.
  • Includes two replacement o-rings and one replacement c-clip.
  • Maximum pressure rating of 40 MPa.
Catalog No. 50-785-425 Supplier Waters Corp Supplier No. WAT037525
The Waters End Connector Kit is designed for use with 3.9 mm or 4.6 mm I.D. cartridges. It allows for the replacement of the cartridge while reusing the end-fittings. This kit has a maximum pressure rating of 40 MPa.
  • Can be used with any 3.9 mm or 4.6 mm I.D. cartridge.
  • Includes two replacement o-rings and one replacement c-clip.
  • Maximum pressure rating of 40 MPa.