Learn More
Microsolv Technology Corporation End Fitting, PEEK, 4.6mm ID, for use with the All-PEEK Column Packing System. 1 EA.
Catalog No. 502271571
Encompass_Preferred
This End Fitting is made for 4.6mm ID All-PEEK Columns with standard HPLC column 10-32 (tubing inlet) screw threads and 1/2"-20 "column tube" screw threads.Designed for use only with our All-PEEK HPLC Column Hardware and not recommended for use with stainless steel columns. Machined from Victrex Brand PEEK which ensures a biocompatibility from the beginning of the packing and column use.
This item is not returnable. View return policy
Catalog No. 50-227-1571 Supplier Microsolv Technology Corporation Supplier No. 49333EF4
This End Fitting is made for 4.6mm ID All-PEEK Columns with standard HPLC column 10-32 (tubing inlet) screw threads and 1/2"-20 "column tube" screw threads.Designed for use only with our All-PEEK HPLC Column Hardware and not recommended for use with stainless steel columns. Machined from Victrex Brand PEEK which ensures a biocompatibility from the beginning of the packing and column use.