Learn More
Electron Microscopy Sciences Microscope Slide End Label SER-16, Standard, 15/32" x 15/16"
SDP
Catalog No. 5029963
Encompass_Preferred
These specialty Microscope Slide Labels and matching End Labels are available in standard (thin) or pathology (tissue high) thickness, and square or round corner (RC). Permanent adhesive holds labels in place during use and long-term storage. Sheet Form Size is 5 1/4" x 8". Standard thin thickness. Square corners.
Use: Standard
Size: 15/32" x 15/16"
Catalog No. 50-299-63 Supplier Electron Microscopy Sciences Supplier No. 7702050
These specialty Microscope Slide Labels and matching End Labels are available in standard (thin) or pathology (tissue high) thickness, and square or round corner (RC). Permanent adhesive holds labels in place during use and long-term storage. Sheet Form Size is 5 1/4" x 8". Standard thin thickness. Square corners.
Use: Standard
Size: 15/32" x 15/16"