Learn More
Electron Microscopy Sciences Microscope Slide End Label SERP-16, Pathology, 15/32" x 15/16"
SDP
Supplier:  Electron Microscopy Sciences 7702055
Encompass_Preferred
These specialty Microscope Slide Labels and matching End Labels are available in standard (thin) or pathology (tissue high) thickness, and square or round corner (RC). Permanent adhesive holds labels in place during use and long-term storage. Sheet Form Size is 5 1/4" x 8". Pathology tissue high thickness. Square corners.
Use: Pathology
Size: 15/32" x 15/16"
Catalog No. 50-299-65