Learn More
Grainger End Mill Kyocera EM 0625 0.6250 Dia
Catalog No. 502763696
Encompass_Preferred
The Kyocera 170GZ2 is a 2 flute, uncoated carbide end mill with a diameter of 0.625 inches and an overall length of 2.5 inches. It has a 0.25 inch shank diameter and is designed for general purpose milling in steels, stainless steels, cast iron, aluminum, and other non-ferrous metals. The end mill features a 30 degree helix angle and is center cutting for plunge milling. According to the product details, it achieves optimal performance at surface speeds from 200 to 300 SFM with feed rates from 0.004 to 0.012 IPR per tooth. The end mill is made in Japan and comes with Kyocera's ZX coating for heat resistance and lubricity.
Catalog No. 50-276-3696 Supplier Grainger Supplier No. 170GZ2
The Kyocera 170GZ2 is a 2 flute, uncoated carbide end mill with a diameter of 0.625 inches and an overall length of 2.5 inches. It has a 0.25 inch shank diameter and is designed for general purpose milling in steels, stainless steels, cast iron, aluminum, and other non-ferrous metals. The end mill features a 30 degree helix angle and is center cutting for plunge milling. According to the product details, it achieves optimal performance at surface speeds from 200 to 300 SFM with feed rates from 0.004 to 0.012 IPR per tooth. The end mill is made in Japan and comes with Kyocera's ZX coating for heat resistance and lubricity.