Learn More
Grainger End Mill Kyocera EM 0750 0.7500 Dia
Catalog No. 502764775
Encompass_Preferred
The Kyocera 170GZ6 is a 1/2" diameter, 4 flute, carbide end mill with a 1-1/2" overall length. It has a TiAIN coating and can be run at speeds up to 8,000 RPM with feed rates up to 80 IPM. This end mill is designed for slotting, profiling, and contouring in steels and cast irons. It features a center cutting design and a 30 degree helix angle. The 170GZ6 has an uncoated solid carbide shank and is made in Japan by Kyocera.
Catalog No. 50-276-4775 Supplier Grainger Supplier No. 170GZ6
The Kyocera 170GZ6 is a 1/2" diameter, 4 flute, carbide end mill with a 1-1/2" overall length. It has a TiAIN coating and can be run at speeds up to 8,000 RPM with feed rates up to 80 IPM. This end mill is designed for slotting, profiling, and contouring in steels and cast irons. It features a center cutting design and a 30 degree helix angle. The 170GZ6 has an uncoated solid carbide shank and is made in Japan by Kyocera.