Learn More
Grainger End Mill Kyocera EM 075009 0.7500 Dia
Catalog No. 502763894
Encompass_Preferred
The Kyocera 170GZ7 is a 2 flute, uncoated carbide end mill with a diameter of 0.25 inches and an overall length of 2 inches. It has a 0.125 inch shank diameter and is designed for high speed machining in aluminum, brass, bronze, plastics and other non-ferrous materials. The end mill features a 30 degree helix angle and can achieve optimal performance at speeds up to 25,000 RPM with 0.002-0.010 inches per tooth feed rates. With its micrograin carbide construction, the 170GZ7 provides high wear resistance and tool life.
Catalog No. 50-276-3894 Supplier Grainger Supplier No. 170GZ7
The Kyocera 170GZ7 is a 2 flute, uncoated carbide end mill with a diameter of 0.25 inches and an overall length of 2 inches. It has a 0.125 inch shank diameter and is designed for high speed machining in aluminum, brass, bronze, plastics and other non-ferrous materials. The end mill features a 30 degree helix angle and can achieve optimal performance at speeds up to 25,000 RPM with 0.002-0.010 inches per tooth feed rates. With its micrograin carbide construction, the 170GZ7 provides high wear resistance and tool life.