Learn More
Grainger End Mill Kyocera EM 100035 100 Dia
Catalog No. 502764781
Encompass_Preferred
The Kyocera End Mill is a high-performance end mill made of micrograin carbide with TiAlN coating for high hardness and heat resistance. It has 4 flutes and a 30 degree helix angle. The end mill comes in diameters ranging from 1/8" to 1/2", with an overall length of 2-1/2". It can achieve surface finishes as fine as 32 microinch Ra. The end mill is designed for high speed machining in steel, cast iron, stainless steel, titanium, nickel alloys, and other hard materials. With its sharp cutting edges and wear resistance, this end mill provides excellent tool life and performance in demanding applications.
Catalog No. 50-276-4781 Supplier Grainger Supplier No. 170HA9
The Kyocera End Mill is a high-performance end mill made of micrograin carbide with TiAlN coating for high hardness and heat resistance. It has 4 flutes and a 30 degree helix angle. The end mill comes in diameters ranging from 1/8" to 1/2", with an overall length of 2-1/2". It can achieve surface finishes as fine as 32 microinch Ra. The end mill is designed for high speed machining in steel, cast iron, stainless steel, titanium, nickel alloys, and other hard materials. With its sharp cutting edges and wear resistance, this end mill provides excellent tool life and performance in demanding applications.