Learn More
Grainger End Mill Kyocera EM 1250 1.2500 Dia
Catalog No. 502764347
Encompass_Preferred
The Kyocera 170HC3 is a 3 flute, high speed steel end mill with a 1/2" shank diameter, 1-1/2" overall length, and 3/4" cutting diameter. It has a TiAIN coating for heat and wear resistance. The end mill features variable helix flutes to reduce chatter and vibration. It is designed for high speed milling in steel, stainless steel, hardened steel, titanium, inconel, waspaloy, nimonics, and cast iron. The end mill provides high metal removal rates and smooth milling action. The 170HC3 is made in Japan by Kyocera, a manufacturer of precision cutting tools.
Catalog No. 50-276-4347 Supplier Grainger Supplier No. 170HC3
The Kyocera 170HC3 is a 3 flute, high speed steel end mill with a 1/2" shank diameter, 1-1/2" overall length, and 3/4" cutting diameter. It has a TiAIN coating for heat and wear resistance. The end mill features variable helix flutes to reduce chatter and vibration. It is designed for high speed milling in steel, stainless steel, hardened steel, titanium, inconel, waspaloy, nimonics, and cast iron. The end mill provides high metal removal rates and smooth milling action. The 170HC3 is made in Japan by Kyocera, a manufacturer of precision cutting tools.