Learn More
Grainger End Mill Kyocera EM 1250AL 1.2500 Dia
Catalog No. 502763829
Encompass_Preferred
The Kyocera 170HC8 is a 1/2" diameter, 4 flute, high speed steel end mill with a 1-1/2" overall length. It has a TiAlN coating for increased hardness and heat resistance. The end mill has a 30 degree helix angle and can be run at speeds up to 325 SFM. It is designed for slotting, profiling, and contouring in steels and cast irons. The end mill features Kyocera's KY2000 coating for high abrasion resistance.
Catalog No. 50-276-3829 Supplier Grainger Supplier No. 170HC8
The Kyocera 170HC8 is a 1/2" diameter, 4 flute, high speed steel end mill with a 1-1/2" overall length. It has a TiAlN coating for increased hardness and heat resistance. The end mill has a 30 degree helix angle and can be run at speeds up to 325 SFM. It is designed for slotting, profiling, and contouring in steels and cast irons. The end mill features Kyocera's KY2000 coating for high abrasion resistance.