Learn More
Grainger End Mill Kyocera EM 1500AL 1.5000 Dia
Catalog No. 502764791
Encompass_Preferred
The Kyocera 170HD3 is a 3 flute, high-speed steel end mill with a 1/2" shank diameter and a 1-1/2" overall length. It has a 3/4" cutting diameter and can be run at speeds up to 325 SFM. This end mill is center cutting and right hand spiral. It is designed for slotting, side milling and contour milling in both ferrous and non-ferrous metals. The 170HD3 features a TiAlN coating for increased tool life and heat resistance.
Catalog No. 50-276-4791 Supplier Grainger Supplier No. 170HD3
The Kyocera 170HD3 is a 3 flute, high-speed steel end mill with a 1/2" shank diameter and a 1-1/2" overall length. It has a 3/4" cutting diameter and can be run at speeds up to 325 SFM. This end mill is center cutting and right hand spiral. It is designed for slotting, side milling and contour milling in both ferrous and non-ferrous metals. The 170HD3 features a TiAlN coating for increased tool life and heat resistance.