Learn More
Grainger End Mill Kyocera MEC 0500S50011 0.5000
Catalog No. 502764758
Encompass_Preferred
The Kyocera 61NV71 is a 4 flute end mill made of micrograin carbide with a TiAIN coating. It has a 1/2" shank diameter, 1-1/2" overall length, and 3/4" cutting diameter. The end mill features variable helix flutes for reduced harmonics and chatter and high positive radial and axial rake angles for easy chip evacuation. Recommended cutting parameters are 12,000-18,000 RPM with feed rates from 0.004-0.012 IPR depending on material. It is well-suited for slotting, profiling, and facing in steels, stainless steels, cast iron, aluminum, and other non-ferrous metals.
Catalog No. 50-276-4758 Supplier Grainger Supplier No. 170HG0
The Kyocera 61NV71 is a 4 flute end mill made of micrograin carbide with a TiAIN coating. It has a 1/2" shank diameter, 1-1/2" overall length, and 3/4" cutting diameter. The end mill features variable helix flutes for reduced harmonics and chatter and high positive radial and axial rake angles for easy chip evacuation. Recommended cutting parameters are 12,000-18,000 RPM with feed rates from 0.004-0.012 IPR depending on material. It is well-suited for slotting, profiling, and facing in steels, stainless steels, cast iron, aluminum, and other non-ferrous metals.