Learn More
Grainger End Mill Kyocera MEC 0750S62511T 0.750
Supplier:  Grainger 170HG2
Encompass_Preferred
The Kyocera 170HG2 is a 2 flute, high speed steel end mill with a 1/2" shank diameter. It has an overall length of 3-1/2" and a cutting diameter of 1/2". The end mill features a 135 degree split point tip and is designed for high feed milling in a variety of materials including steels, cast iron, aluminum, copper, brass, plastics and composites. With its high speed steel construction, this end mill provides good wear resistance and tool life. The 170HG2 is well-suited for slotting, side milling and contouring operations.
Catalog No. 50-276-3882