Learn More
Grainger End Mill Kyocera MEC 10S1011 10mm D
Catalog No. 502764754
Encompass_Preferred
The Kyocera 170HL4 is a 4 flute, high speed steel end mill with a length of cut of 1-1/2". It has a 1/2" shank diameter and an overall length of 3-1/2". The end mill has a TiAIN coating for increased tool life and heat resistance. It is designed for high feed milling of steel, stainless steel, and cast iron. The end mill provides high metal removal rates in slotting, plunging and ramping applications.
Catalog No. 50-276-4754 Supplier Grainger Supplier No. 170HL4
The Kyocera 170HL4 is a 4 flute, high speed steel end mill with a length of cut of 1-1/2". It has a 1/2" shank diameter and an overall length of 3-1/2". The end mill has a TiAIN coating for increased tool life and heat resistance. It is designed for high feed milling of steel, stainless steel, and cast iron. The end mill provides high metal removal rates in slotting, plunging and ramping applications.