Learn More
Grainger End Mill Kyocera MEC 1500S12511T 1.500
Catalog No. 502764676
Encompass_Preferred
The Kyocera 170HH3 is a 1/2" diameter, 4 flute, high helix end mill made of micrograin carbide with TiAlN coating. It has a 1-1/2" LOC and can be run at speeds up to 325 SFM. The end mill features variable helix flutes to reduce harmonics and chatter and is designed for slotting, profiling, and contouring in steels, stainless steels, cast iron, aluminum, and other non-ferrous metals. It has a center cutting design allowing plunge cuts and is made in Japan to tight tolerances. The end mill comes with a 30 day warranty against manufacturing defects.
Catalog No. 50-276-4676 Supplier Grainger Supplier No. 170HH3
The Kyocera 170HH3 is a 1/2" diameter, 4 flute, high helix end mill made of micrograin carbide with TiAlN coating. It has a 1-1/2" LOC and can be run at speeds up to 325 SFM. The end mill features variable helix flutes to reduce harmonics and chatter and is designed for slotting, profiling, and contouring in steels, stainless steels, cast iron, aluminum, and other non-ferrous metals. It has a center cutting design allowing plunge cuts and is made in Japan to tight tolerances. The end mill comes with a 30 day warranty against manufacturing defects.