Learn More
Grainger End Mill Kyocera MEC 16S1611T 16mm
Catalog No. 502764756
Encompass_Preferred
The Kyocera 170HL3 is a 1/2" diameter, 4 flute, high speed steel end mill with a 3" overall length. It has a TiAlN coating for increased hardness and heat resistance. The end mill has a 30 degree helix angle and is center cutting for plunge cuts. It is designed for slotting, profiling, and contouring in steels, cast iron, aluminum, and other non-ferrous metals at speeds up to 325 SFM. The end mill has a tolerance of +/- .0005" and comes with a 30 day warranty from the manufacturer.
Catalog No. 50-276-4756 Supplier Grainger Supplier No. 170HL3
The Kyocera 170HL3 is a 1/2" diameter, 4 flute, high speed steel end mill with a 3" overall length. It has a TiAlN coating for increased hardness and heat resistance. The end mill has a 30 degree helix angle and is center cutting for plunge cuts. It is designed for slotting, profiling, and contouring in steels, cast iron, aluminum, and other non-ferrous metals at speeds up to 325 SFM. The end mill has a tolerance of +/- .0005" and comes with a 30 day warranty from the manufacturer.