Learn More
Grainger End Mill Kyocera MEC 17S1611T 17mm
Catalog No. 502764356
Encompass_Preferred
The Kyocera 170HP9 is a 1/2" diameter, 4 flute, high performance cobalt end mill with a 1-1/2" overall length. It has a TiAIN coating and can machine a variety of materials including hardened steels up to 65HRC, high temp alloys, titanium alloys, nickel based alloys, and stainless steels. The end mill features variable helix flutes to reduce harmonics and provide excellent chip control. Recommended cutting parameters are 12,000-18,000 RPM with feed rates from 0.004-0.012 IPR depending on the material being machined.
Catalog No. 50-276-4356 Supplier Grainger Supplier No. 170HP9
The Kyocera 170HP9 is a 1/2" diameter, 4 flute, high performance cobalt end mill with a 1-1/2" overall length. It has a TiAIN coating and can machine a variety of materials including hardened steels up to 65HRC, high temp alloys, titanium alloys, nickel based alloys, and stainless steels. The end mill features variable helix flutes to reduce harmonics and provide excellent chip control. Recommended cutting parameters are 12,000-18,000 RPM with feed rates from 0.004-0.012 IPR depending on the material being machined.