Learn More
Grainger End Mill Kyocera MEC 25S2521017 25m
Catalog No. 502764264
Encompass_Preferred
The Kyocera 170HP7 is a 1/2" diameter, 4 flute, high performance cobalt end mill with a 1-1/2" overall length. It has a TiAIN coating and can be run at speeds up to 325 SFM. The end mill features variable helix flutes to reduce harmonics and provide a smooth cut. It is designed for high temp alloys, hardened steels, titanium and other exotics. The end mill is made in Japan.
Catalog No. 50-276-4264 Supplier Grainger Supplier No. 170HP7
The Kyocera 170HP7 is a 1/2" diameter, 4 flute, high performance cobalt end mill with a 1-1/2" overall length. It has a TiAIN coating and can be run at speeds up to 325 SFM. The end mill features variable helix flutes to reduce harmonics and provide a smooth cut. It is designed for high temp alloys, hardened steels, titanium and other exotics. The end mill is made in Japan.