Learn More
Grainger End Mill Kyocera MEC 32S2511T 32mm
Catalog No. 502764751
Encompass_Preferred
The Kyocera 170HM0 is a 1/2" diameter, 4 flute, high speed steel end mill with a 1-1/2" overall length. It has a TiAlN coating for increased hardness and heat resistance. The end mill has a 30 degree helix angle and is center cutting for plunge cuts. It is designed for slotting, profiling, and contouring in steels, cast iron, aluminum, and other non-ferrous metals at speeds up to 325 SFM. The end mill has a tolerance of +/- .0005" and comes with a 30 day warranty from the manufacturer.
Catalog No. 50-276-4751 Supplier Grainger Supplier No. 170HM0
The Kyocera 170HM0 is a 1/2" diameter, 4 flute, high speed steel end mill with a 1-1/2" overall length. It has a TiAlN coating for increased hardness and heat resistance. The end mill has a 30 degree helix angle and is center cutting for plunge cuts. It is designed for slotting, profiling, and contouring in steels, cast iron, aluminum, and other non-ferrous metals at speeds up to 325 SFM. The end mill has a tolerance of +/- .0005" and comes with a 30 day warranty from the manufacturer.