Learn More
Grainger End Mill Kyocera MECX 0375S375071T 0.3
Catalog No. 502764765
Encompass_Preferred
The Kyocera MECX 0375S375071T is a 3/4" diameter, 3/8" shank, 4 flute, high speed steel end mill. It has a TiAlN coating for increased hardness and heat resistance. The overall length is 3-3/4" and it has a 1/2" LOC. This end mill is designed for slotting, profiling, contouring, and plunging on materials like alloy steels, tool steels, hardened steels, titanium alloys, nickel based alloys, and stainless steels. It has a tolerance of +/- .005" and concentricity of .002" TIR. The end mill has a 30 degree helix angle.
Catalog No. 50-276-4765 Supplier Grainger Supplier No. 170HM4
The Kyocera MECX 0375S375071T is a 3/4" diameter, 3/8" shank, 4 flute, high speed steel end mill. It has a TiAlN coating for increased hardness and heat resistance. The overall length is 3-3/4" and it has a 1/2" LOC. This end mill is designed for slotting, profiling, contouring, and plunging on materials like alloy steels, tool steels, hardened steels, titanium alloys, nickel based alloys, and stainless steels. It has a tolerance of +/- .005" and concentricity of .002" TIR. The end mill has a 30 degree helix angle.