Learn More
Grainger End Mill Kyocera MECX 0625S625073T 0.6
Supplier:  Grainger 170HM6
Encompass_Preferred
The Kyocera 170HM6 is a 1/2" diameter, 4 flute, high speed steel end mill with a 1-1/2" overall length. It has a TiAlN coating for increased hardness and heat resistance. The end mill has a 30 degree helix angle and can be run at speeds up to 325 SFM. It is designed for slotting, profiling, contouring, and facing in a variety of materials including steels, cast iron, aluminum, copper alloys, plastics and composites. The end mill has a tolerance of +/- .0005" and comes with a 30 day warranty against manufacturer defects.
Catalog No. 50-276-3886