Learn More
Grainger End Mill Kyocera MECX 1000S100077T 1.0
Catalog No. 502764350
Encompass_Preferred
The Kyocera 170HN3 is a 1/2" diameter, 4 flute, high speed steel end mill with a 3/4" overall length. It has a TiAlN coating for increased hardness and heat resistance. The end mill has a 30 degree helix angle and can be run at speeds up to 325 SFM. It is designed for slotting, profiling, and contouring in steels, cast iron, aluminum, and other non-ferrous metals. The end mill features a center cutting design for plunge cuts and slotting.
Catalog No. 50-276-4350 Supplier Grainger Supplier No. 170HN3
The Kyocera 170HN3 is a 1/2" diameter, 4 flute, high speed steel end mill with a 3/4" overall length. It has a TiAlN coating for increased hardness and heat resistance. The end mill has a 30 degree helix angle and can be run at speeds up to 325 SFM. It is designed for slotting, profiling, and contouring in steels, cast iron, aluminum, and other non-ferrous metals. The end mill features a center cutting design for plunge cuts and slotting.