Learn More
Grainger End Mill Kyocera MECX 16S16074T 16m
Catalog No. 502764832
Encompass_Preferred
The Kyocera 170HP2 is a 1/2" diameter, 4 flute, high performance cobalt end mill with a 1-1/2" overall length. It has a TiAIN coating and can machine a variety of materials including hardened steels up to 65 HRC, high temp alloys, titanium alloys, nickel based alloys, and stainless steels. The end mill features variable helix flutes to reduce harmonics and provide excellent chip control. Recommended cutting parameters are 12,000-18,000 RPM with feed rates from 0.004-0.012 IPR depending on the material being machined.
Catalog No. 50-276-4832 Supplier Grainger Supplier No. 170HP2
The Kyocera 170HP2 is a 1/2" diameter, 4 flute, high performance cobalt end mill with a 1-1/2" overall length. It has a TiAIN coating and can machine a variety of materials including hardened steels up to 65 HRC, high temp alloys, titanium alloys, nickel based alloys, and stainless steels. The end mill features variable helix flutes to reduce harmonics and provide excellent chip control. Recommended cutting parameters are 12,000-18,000 RPM with feed rates from 0.004-0.012 IPR depending on the material being machined.