Learn More
Grainger End Mill Kyocera MECX 32S32078T 32m
Catalog No. 502763830
Encompass_Preferred
The Kyocera 170HP6 is a 1/2" diameter, 4 flute, high performance cobalt end mill with a 1-1/2" overall length. It has a TiAIN coating and can machine a variety of materials including hardened steels up to 60 HRC, high temp alloys, titanium alloys, nickel based alloys, and stainless steels. The end mill has a center cutting design and can be run at speeds up to 325 SFM. It is made in Japan by Kyocera and is ideal for high performance milling applications.
Catalog No. 50-276-3830 Supplier Grainger Supplier No. 170HP6
The Kyocera 170HP6 is a 1/2" diameter, 4 flute, high performance cobalt end mill with a 1-1/2" overall length. It has a TiAIN coating and can machine a variety of materials including hardened steels up to 60 HRC, high temp alloys, titanium alloys, nickel based alloys, and stainless steels. The end mill has a center cutting design and can be run at speeds up to 325 SFM. It is made in Japan by Kyocera and is ideal for high performance milling applications.